Anti-CBL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-3089R-A647
BOSSBS-3089R-A647EA 921 EUR InStock
BOSSBS-3089R-A647
Anti-CBL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
The proto oncogene c CBL was initially identified as the cellular homologue of v CBL oncogene that induces pre B cell lymphomas and myeloid leukemias in mice. In more recent studies CBL has been shown to be a negative regulator of tyrosine kinase signaling. The ubiquitin ligase activity of CBL leads to the degradation of tyrosine kinases, thus attenuating the signal of receptors. Targets of CBL include activated protein tyrosine kinases belonging to the Src and Syk/Zap 70 families. An additional mechanism to attenuate receptor signaling is thought to be achieved by CBL’s interaction withMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité