Anti-TNK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-3145R-FITC
BOSSBS-3145R-FITCEA 929 EUR InStock
BOSSBS-3145R-FITC
Anti-TNK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
This gene encodes a member of the O class of winged helix/forkhead transcription factor family. Proteins encoded by this class are regulated by factors involved in growth and differentiation indicating they play a role in these processes. A translocation involving this gene on chromosome X and the homolog of the Drosophila trithorax gene, encoding a DNA binding protein, located on chromosome 11 is associated with leukemia. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2010]
Recommended Dilutions: IF(IHC-P): 1:50-200
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-3145R-FITC
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité