Anti-PTK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-3159R-A555
BOSSBS-3159R-A555EA 898 EUR InStock
BOSSBS-3159R-A555
Anti-PTK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. Several transcriptMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-3159R-A555
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité