Anti-CK17 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-3239R-CY5.5
BOSSBS-3239R-CY5.5EA 929 EUR InStock
BOSSBS-3239R-CY5.5
Anti-CK17 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. Unlike its related family members, this smallest known acidic cytokeratin is not paired with a basic cytokeratin in epithelial cells. It is specifically expressed in the periderm, the transiently superficial layer that envelopes the developing epidermis.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité