Anti-PDGFRA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-3318R-A555
BOSSBS-3318R-A555EA 921 EUR InStock
BOSSBS-3318R-A555
Anti-PDGFRA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
The PDGF Receptor Type A (Alpha platelet-derived growth factor receptor precursor, CD140a antigen), a 170kD protein, binds all three isoforms of PDGF with high affinity whereas the PDGF Receptor Type B, a 190kD protein, appears to bind only the PDGF BB homodimer with high affinity. Both receptors are transmembranous, ligand activated protein tyrosine kinases, which phosphorylate a number of important signal transduction proteins, which are bound with differential affinities via SH2 domains. The response of any given cell to PDGF will depend on the types of receptors displayed on the surfaceMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité