Anti-PKM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-3334R-A647
BOSSBS-3334R-A647EA 921 EUR InStock
BOSSBS-3334R-A647
Anti-PKM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Glycolytic enzyme that catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate (PEP) to ADP, generating ATP. Stimulates POU5F1-mediated transcriptional activation. Plays a general role in caspase independent cell death of tumor cells. The ratio betwween the highly active tetrameric form and nearly inactive dimeric form determines whether glucose carbons are channeled to biosynthetic processes or used for glycolytic ATP production. The transition between the 2 forms contributes to the control of glycolysis and is important for tumor cell proliferation and survival.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité