Anti-SIRT1 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-3393R-CY5.5
BOSSBS-3393R-CY5.5EA 929 EUR InStock
BOSSBS-3393R-CY5.5
Anti-SIRT1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité