Anti-PTK2B Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-3401R-CY5.5
BOSSBS-3401R-CY5.5EA 921 EUR InStock
BOSSBS-3401R-CY5.5
Anti-PTK2B Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
PYK2 is involved in calcium induced regulation of ion channel and activation of the map kinase signaling pathway. PKY2 may represent an important signaling intermediate between neuropeptide activated receptors or neurotransmitters that increase calcium flux and the downstream signals that regulate neuronal activity. Interacts with the SH2 domain of Grb2. May phosphorylate the voltage gated potassium channel protein Kv1.2. PYK2 activation is highly correlated with the stimulation of c-Jun N-terminal kinase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTK2B
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité