Anti-TRIM28 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-3581R-CY5.5
BOSSBS-3581R-CY5.5EA 900 EUR InStock
BOSSBS-3581R-CY5.5
Anti-TRIM28 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Nuclear corepressor for KRAB domain-containing zinc finger proteins (KRAB-ZFPs). Mediates gene silencing by recruiting CHD3, a subunit of the nucleosome remodeling and deacetylation (NuRD) complex, and SETDB1 (which specifically methylates histone H3 at 'Lys-9' (H3K9me)) to the promoter regions of KRAB target genes. Enhances transcriptional repression by coordinating the increase in H3K9me, the decrease in histone H3 'Lys-9 and 'Lys-14' acetylation (H3K9ac and H3K14ac, respectively) and the disposition of HP1 proteins to silence gene expression. Recruitment of SETDB1 inducesMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité