Anti-Histone H2B Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-3652R-CY3
BOSSBS-3652R-CY3EA 900 EUR InStock
BOSSBS-3652R-CY3
Anti-Histone H2B Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H2B family. Transcripts from this gene lack polyA tails; instead, they contain aMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité