Anti-cGMP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-3892R-A750
BOSSBS-3892R-A750EA 890 EUR InStock
BOSSBS-3892R-A750
Anti-cGMP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Cyclic guanosine monophosphate (cGMP) serves as a second messenger in a manner similar to that observed with cAMP. Peptide hormones, such as the natriuretic factors, activate receptors that are associated with membrane-bound guanylate cyclase (GC). Receptor activation of GC leads to the conversion of GTP to cGMP. Nitric oxide (NO) also stimulates cGMP production by activating soluble GC, perhaps by binding to the heme moiety of the enzyme. Similar to cAMP, cGMP mediates most of its intracellular effects through the activation of specific cGMP dependent protein kinases (PKG).
Type: Primary
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité