Anti-CUBN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-3901R-A647
BOSSBS-3901R-A647EA 893 EUR InStock
BOSSBS-3901R-A647
Anti-CUBN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Cotransporter which plays a role in lipoprotein, vitamin and iron metabolism, by facilitating their uptake. Binds to ALB, MB, Kappa and lambda-light chains, TF, hemoglobin, GC, SCGB1A1, APOA1, high density lipoprotein, and the GIF-cobalamin complex. The binding of all ligands requires calcium. Serves as important transporter in several absorptive epithelia, including intestine, renal proximal tubules and embryonic yolk sac. Interaction with LRP2 mediates its trafficking throughout vesicles and facilitates the uptake of specific ligands like GC, hemoglobin, ALB, TF and SCGB1A1. InteractionMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité