Anti-HLA-DPB1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-4107R-A680
BOSSBS-4107R-A680EA 890 EUR InStock
BOSSBS-4107R-A680
Anti-HLA-DPB1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Binds peptides derived from antigens that access the endocytic route of antigen presenting cells (APC) and presents them on the cell surface for recognition by the CD4 T-cells. The peptide binding cleft accommodates peptides of 10-30 residues. The peptides presented by MHC class II molecules are generated mostly by degradation of proteins that access the endocytic route, where they are processed by lysosomal proteases and other hydrolases. Exogenous antigens that have been endocytosed by the APC are thus readily available for presentation via MHC II molecules, and for this reason thisMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité