Anti-MEF2D Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-4136R-A488
BOSSBS-4136R-A488EA 893 EUR InStock
BOSSBS-4136R-A488
Anti-MEF2D Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Transcriptional activator which binds specifically to the MEF2 element, 5'-YTA[AT](4)TAR-3', found in numerous muscle-specific, growth factor- and stress-induced genes. Mediates cellular functions not only in skeletal and cardiac muscle development, but also in neuronal differentiation and survival. Plays diverse roles in the control of cell growth, survival and apoptosis via p38 MAPK signaling in muscle-specific and/or growth factor-related transcription. Plays a critical role in the regulation of neuronal apoptosis (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité