Anti-EGF Mouse Polyclonal Antibody
Fournisseur: Bioss
BS-4568M
BOSSBS-4568MEA 1090 EUR InStock
BOSSBS-4568M
Anti-EGF Mouse Polyclonal Antibody
Anticorps
Epidermal growth factor (EGF) is an acid- and heat-stable 53 amino acid protein originally found in rodents and humans. It has been shown to be a potent mitogen for a variety of cell types both in vivo and in vitro. EGF binds to the EGF receptor on the surface of cells and mediates intrinsic phosphorylation of the receptor on tyrosine residues. It has been detected in nearly all body fluids, such as urine (urogastrone), saliva, milk and platelet-rich plasma. EGF, TGF?and vaccinia virus growth factor exhibit 30-40% amino acid homology. Several additional members of the EGF/TGF family haveMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité