Anti-Gibberellins Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-4606R
BOSSBS-4606REA 725 EUR InStock
BOSSBS-4606R
Anti-Gibberellins Rabbit Polyclonal Antibody
Anticorps
Gibberillic acids (GA) are important plant growth hormones that promote plant cell growth and elongation. Gebberillins promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. The gibberellins are also involved in processes such as gravitropism, tensioning and floral display.Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media. The specific actions of A3 and A4 or combinations of Gebberellins should be determined in specific plant applications.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité