Anti-RBL2 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-4984R
BOSSBS-4984REA 725 EUR InStock
BOSSBS-4984R
Anti-RBL2 Rabbit Polyclonal Antibody
Anticorps
p130 is related both in structure and function to the retinoblastoma tumor suppressor protein and p107 (collectively known as pocket proteins) and is known to regulate the activity of E2F transcription factors. E2F transcription factors regulate the expression of a number of genes important in cell proliferation, particularly those involved in the progression through G1 and into the S phase of the cell cycle. Binding of p130 converts E2F transcription factors from transcriptional activators to transcriptional repressors.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité