Anti-AGER Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-4999R-A750
BOSSBS-4999R-A750EA 899 EUR InStock
BOSSBS-4999R-A750
Anti-AGER Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Mediates interactions of advanced glycosylation end products (AGE). These are nonenzymatically glycosylated proteins which accumulate in vascular tissue in aging and at an accelerated rate in diabetes. Acts as a mediator of both acute and chronic vascular inflammation in conditions such as atherosclerosis and in particular as a complication of diabetes. AGE/RAGE signaling plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes. Interaction with S100A12 on endothelium, mononuclear phagocytes, andMore Product Information
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
VIA 100 µl BOSSBS-4999R-A750
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité