Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-BRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fournisseur: Bioss
BS-5124R-A647
BOSSBS-5124R-A647EA 893 EUR InStock
BOSSBS-5124R-A647
Anti-BRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
HSPC300 (hematopoietic stem cell protein 300) is also known as probable protein BRICK1 or C3orf10 (chromosome 3 open reading frame 10) and is a 75 amino acid protein that is expressed as two isoforms and localizes to both the cytoplasm and the cytoskeleton. HSPC300 is thought to regulate cytoskeletal organization and Actin polymerization. Free HSPC300 exists as homotrimers prior to its incorporation into the WAVE complex. The WAVE complex includes five proteins, one of which is HSPC300, that regulate the ARC (Arp2/3 complex) which is responsible for Actin nucleation and is Rac 1-dependent. Because HSPC300 is a highly conserved subunit of the WAVE complex across many species, it is thought to have the same or similar functions in many different organisms. In Drosophila, the WAVE/ARC pathway may affect the development of the nervous system. HSPC300 is thought to localize to axons of the central nervous system of Drosophila embryos and thus may also be involved in axonogenesis. In addition, HSPC300 is thought to be necessary for synaptic morphogenesis by motoneurons. In mice, the knockout of the WAVE complex leads to learning and memory defects, and it is therefore hypothesized that HSPC300 may also be involved in cognitive functions. Genetic depletion of HSPC300 results in cytoskeletal abnormalities and prevents cytokinesis of cells, suggesting that decreased levels of HSPC300 may be associated with tumor suppression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité