Anti-AXL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-5180R-A647
BOSSBS-5180R-A647EA 915 EUR InStock
BOSSBS-5180R-A647
Anti-AXL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factor GAS6 and which is thus regulating many physiological processes including cell survival, cell proliferation, migration and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of AXL. Following activation by ligand, ALX binds and induces tyrosine phosphorylation of PI3-kinase subunits PIK3R1, PIK3R2 and PIK3R3; but also GRB2, PLCG1, LCK and PTPN11. Other downstream substrate candidates for AXL are CBL, NCK2, SOCS1 and TENC1.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité