Anti-DAXX Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-5292R-A647
BOSSBS-5292R-A647EA 922 EUR InStock
BOSSBS-5292R-A647
Anti-DAXX Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. Cell death signals are transduced by death domain (DD) containing adapter molecules and members of the ICE/CED3 protease family. A novel DD containing molecule was recently cloned from mouse, human and monkey and designated Daxx. Daxx is a death domain containing important intermediate in the Fas mediated apoptosis.More Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-5292R-A647
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité