Anti-DAXX Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-5292R-CY7
BOSSBS-5292R-CY7EA 930 EUR InStock
BOSSBS-5292R-CY7
Anti-DAXX Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. Cell death signals are transduced by death domain (DD) containing adapter molecules and members of the ICE/CED3 protease family. A novel DD containing molecule was recently cloned from mouse, human and monkey and designated Daxx. Daxx is a death domain containing important intermediate in the Fas mediated apoptosis.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité