Anti-HSP70 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-5362R-CY5
BOSSBS-5362R-CY5EA 930 EUR InStock
BOSSBS-5362R-CY5
Anti-HSP70 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
RecommendedMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité