Anti-GIT1 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-5373R-CY3
BOSSBS-5373R-CY3EA 930 EUR InStock
BOSSBS-5373R-CY3
Anti-GIT1 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Heterotrimeric G protein-mediated signal transduction is a dynamically regulated process with the intensity of signal decreasing over time despite the continued presence of the agonist (1,2). G protein-coupled receptor kinases (GRKs) are activated by activated G protein-coupled receptors, and they function to phosphorylate and inactivate cell surface receptors in the heterotrimeric G protein signaling cascade (3,4). GIT1 (for GRK-interactor 1) and GIT2 are GTPase-activating proteins (GAP) for members of the ADP ribosylation factor (ARF) family of small GTP-binding proteins, which areMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité