Anti-IL1R1 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-5394R-CY5.5
BOSSBS-5394R-CY5.5EA 930 EUR InStock
BOSSBS-5394R-CY5.5
Anti-IL1R1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Receptor for IL1A, IL1B and IL1RN. After binding to interleukin-1 associates with the corecptor IL1RAP to form the high affinity interleukin-1 receptor complex which mediates interleukin-1-dependent activation of NF-kappa-B, MAPK and other pathways. Signaling involves the recruitment of adapter molecules such as TOLLIP, MYD88, and IRAK1 or IRAK2 via the respective TIR domains of the receptor/coreceptor subunits. Binds ligands with comparable affinity and binding of antagonist IL1RN prevents association with IL1RAP to form a signaling complex.
Recommended Dilutions: IF(IHC-P): 1:50-200
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité