Anti-MAPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-5417R-A488
BOSSBS-5417R-A488EA 922 EUR InStock
BOSSBS-5417R-A488
Anti-MAPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Tau proteins are important Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by tau localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization. TauMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité