Anti-PP1C gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-5422R-A680
BOSSBS-5422R-A680EA 890 EUR InStock
BOSSBS-5422R-A680
Anti-PP1C gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. The protein is involved in regulation of ionic conductances and long term synaptic plasticity. It may play an important role in dephosphorylating substrates such as the postsynaptic density associated Ca (2+)/calmodulin dependent protein kinase II.PP1 comprises a catalytic subunit, PPP1CA, PPP1CB or PPP1CC (PP1C gamma), which is folded into its native form by inhibitor 2 and glycogen synthetase kinase 3, and then complexed to oneMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité