Anti-TAB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-5424R-A488
BOSSBS-5424R-A488EA 890 EUR InStock
BOSSBS-5424R-A488
Anti-TAB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
The product of this gene functions in the NF-kappaB signal transduction pathway. The encoded protein, and the similar and functionally redundant protein MAP3K7IP2/TAB2, forms a ternary complex with the protein kinase MAP3K7/TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent MAP3K7/TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. The human genome contains a related pseudogene. Alternatively spliced transcript variants have been described, but their biologicalMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité