Anti-MAPK3 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-5497R-CY3
BOSSBS-5497R-CY3EA 930 EUR InStock
BOSSBS-5497R-CY3
Anti-MAPK3 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variantsMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité