Anti-YWHAE Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-5590R-A488
BOSSBS-5590R-A488EA 886 EUR InStock
BOSSBS-5590R-A488
Anti-YWHAE Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathway. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: YWHAE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité