Anti-TRIM25 Ser100 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-5615R-A750
BOSSBS-5615R-A750EA 921 EUR InStock
BOSSBS-5615R-A750
Anti-TRIM25 Ser100 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains, a RING, a B box type 1 and a B box type 2, and a coiled coil region. The protein localises to the cytoplasm. The presence of potential DNA binding and dimerization transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to oestrogen, and it is thought to mediate oestrogen actions in breast cancer as a primary response gene.
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
VIA 100 µl BOSSBS-5615R-A750
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité