Anti-MAD2L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-5731R-FITC
BOSSBS-5731R-FITCEA 900 EUR InStock
BOSSBS-5731R-FITC
Anti-MAD2L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Mad2L1 is required for the execution of the mitotic checkpoint which monitors the process of kinetochore spindle attachment and delays the onset of anaphase when this process is not complete. It inhibits the activity of the anaphase promoting complex by sequestering CDC20 until all chromosomes are aligned at the metaphase plate.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité