Anti-ETV7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-5965R-A680
BOSSBS-5965R-A680EA 890 EUR InStock
BOSSBS-5965R-A680
Anti-ETV7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
The protein encoded by this gene belongs to the ETS family of transcription factors, which is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well. This protein is predominantly expressed in hematopoietic tissues. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene (PMID:11108721).[provided by RefSeq, May 2011].
Type: Primary
Antigen: ETV7
Clonality: Polyclonal
Clone:
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité