Anti-APEH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-5983R-A488
BOSSBS-5983R-A488EA 893 EUR InStock
BOSSBS-5983R-A488
Anti-APEH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
AARE (Acylamino-acid-releasing enzyme) is also known as Acyl-peptide hydrolase. It catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. It can play an important role in destroying oxidatively damaged proteins in living cells. Deletions of this gene locus corresponding to the protein are found in various types of carcinomas, including small cell lung carcinoma and renal cell carcinoma.
Recommended Dilutions: IF(IHC-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité