Anti-AIM2 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-5986R-CY5
BOSSBS-5986R-CY5EA 900 EUR InStock
BOSSBS-5986R-CY5
Anti-AIM2 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
Involved in innate immune response by recognizing cytosolic double-stranded DNA and inducing caspase-1-activating inflammasome formation in macrophages. Upon binding to DNA is thought to undergo oligomerization and to associate with PYCARD initiating the recruitment of caspase-1 precusrsor and processing of interleukin-1 beta and interleukin-18. Detects cytosolic dsDNA of viral and bacterial origin in a non-sequence-specific manner. Can also trigger PYCARD-dependent, caspase-1-independent cell death that involves caspase-8.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-2More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité