Anti-CPEB1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-6037R-A750
BOSSBS-6037R-A750EA 893 EUR InStock
BOSSBS-6037R-A750
Anti-CPEB1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Sequence-specific RNA-binding protein that regulates mRNA cytoplasmic polyadenylation and translation initiation during oocyte maturation, early development and at postsynapse sites of neurons. Binds to the cytoplasmic polyadenylation element (CPE), an uridine-rich sequence element (consensus sequence 5'-UUUUUAU-3') within the mRNA 3'-UTR. In absence of phosphorylation and in association with TACC3 is also involved as a repressor of translation of CPE-containing mRNA; a repression that is relieved by phosphorylation or degradation (By similarity). Involved in the transport of CPE-containingMore Product Information
Collapse All
Type de conditionnement Cdt Numéro de catalogue VWR Quantité
VIA 100 µl BOSSBS-6037R-A750
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité