Anti-LLGL1 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-6070R-CY5
BOSSBS-6070R-CY5EA 930 EUR InStock
BOSSBS-6070R-CY5
Anti-LLGL1 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité