Anti-IQGAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-6171R-A350
BOSSBS-6171R-A350EA 893 EUR InStock
BOSSBS-6171R-A350
Anti-IQGAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member has a perinuclear localization and is an inositol 1,3,4,5-tetrakisphosphate-binding protein; a compound suggested to function as a secondMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité