Anti-SETD7 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-6191R
BOSSBS-6191REA 725 EUR InStock
BOSSBS-6191R
Anti-SETD7 Rabbit Polyclonal Antibody
Anticorps
Histone methyltransferase that specifically monomethylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates 'Lys-189' of TAF10, leading to increase the affinity ofMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 μg 100 µl BOSSBS-6191R
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité