Anti-DOK3 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-6234R-CY7
BOSSBS-6234R-CY7EA 900 EUR InStock
BOSSBS-6234R-CY7
Anti-DOK3 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate Abl functionDOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate Abl function. There are 4 isoforms generated byMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité