Anti-SH3KBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-6239R-A647
BOSSBS-6239R-A647EA 893 EUR InStock
BOSSBS-6239R-A647
Anti-SH3KBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
SH3KBP1 belongs to the CIN85/CMS family of adaptor molecules, characterized by containing three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The different members of the family orchestrate a network involved in dowregulation and degradation of recpetor tyrosine kinases. SH3KBP1 is involved in regulating diverse signal transduction pathways. Involved in the regulation of endocytosis and lysosomal degradation of ligand-induced receptor tyrosine kinases, including EGFR and MET/hepatocyte growth factor receptor, through a association with CBL andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité