Anti-TM4SF1 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-6242R-CY7
BOSSBS-6242R-CY7EA 893 EUR InStock
BOSSBS-6242R-CY7
Anti-TM4SF1 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface antigen and is highly expressed in different carcinomas. [provided by RefSeq, Jul 2008]
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TM4More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité