Anti-NEK3 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-6261R
BOSSBS-6261REA 725 EUR InStock
BOSSBS-6261R
Anti-NEK3 Rabbit Polyclonal Antibody
Anticorps
This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Mar2009].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité