Anti-Lin-28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-6446R-A680
BOSSBS-6446R-A680EA 893 EUR InStock
BOSSBS-6446R-A680
Anti-Lin-28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Acts as a suppressor of microRNA (miRNA) biogenesis by specifically binding the precursor let-7 (pre-let-7), a miRNA precursor. Acts by binding pre-let-7 and recruiting ZCCHC11/TUT4 uridylyltransferase, leading to the terminal uridylation of pre-let-7. Uridylated pre-let-7 miRNAs fail to be processed by Dicer and undergo degradation. Specifically recognises the 5'-GGAG-3' motif in the terminal loop of pre-let-7. Also recognises and binds non pre-let-7 pre-miRNAs that contain the 5'-GGAG-3' motif in the terminal loop, leading to their terminal uridylation and subsequent degradation. MediatesMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité