Anti-NLRP9 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-6867R-CY7
BOSSBS-6867R-CY7EA 893 EUR InStock
BOSSBS-6867R-CY7
Anti-NLRP9 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The protein encoded by this gene belongs to the NALP protein family. Members of the NALP protein family typically contain a NACHT domain, a NACHT-associated domain (NAD), a C-terminal leucine-rich repeat (LRR) region, and an N-terminal pyrin domain (PYD). This protein may play a regulatory role in the innate immune system as similar family members belong to the signal-induced multiprotein complex, the inflammasome, that activates the pro-inflammatory caspases, caspase-1 and caspase-5. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NLRP9
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité