Anti-FAM175A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-6921R-HRP
BOSSBS-6921R-HRPEA 900 EUR InStock
BOSSBS-6921R-HRP
Anti-FAM175A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Component of the BRCA1-A complex, a complex that specifically recognizes 'Lys-63'-linked ubiquitinated histones H2A and H2AX at DNA lesions sites, leading to target the BRCA1-BARD1 heterodimer to sites of DNA damage at double-strand breaks (DSBs). The BRCA1-A complex also possesses deubiquitinase activity that specifically removes 'Lys-63'-linked ubiquitin on histones H2A and H2AX. In the BRCA1-A complex, it acts as a central scaffold protein that assembles the various components of the BRCA1-A complex and mediates the recruitment of BRCA1.
Recommended Dilutions: Western Blot: 1:100-1000;More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité