Anti-CCDC51 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-6925R-CY3
BOSSBS-6925R-CY3EA 900 EUR InStock
BOSSBS-6925R-CY3
Anti-CCDC51 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
The coiled-coil domain is a structural motif found in proteins that are involved in a diverse array of biological functions such as the regulation of gene expression, cell division, membrane fusion and drug extrusion and delivery. CCDC51 (coiled-coil domain containing 51) is a 411 amino acid multi-pass membrane protein that exists as two alternatively spliced isoforms. The gene encoding CCDC51 maps to human chromosome 3, which is made up of about 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci onMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-6925R-CY3
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité