Anti-PARP3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-7108R-A488
BOSSBS-7108R-A488EA 893 EUR InStock
BOSSBS-7108R-A488
Anti-PARP3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks. May link the DNA damage surveillance network to the mitotic fidelity checkpoint. Negatively influences the G1/S cell cycle progression without interfering with centrosome duplication. Binds DNA. May be involved in the regulation of PRC2 and PRC3 complex-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité