Anti-MHC class 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-7335R-A680
BOSSBS-7335R-A680EA 893 EUR InStock
BOSSBS-7335R-A680
Anti-MHC class 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
MHC Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. MHC class I antigens are heterodimers consisting of one alpha chain (44kDa) with beta 2 microglobulin (11.5 kDa). The antigen is expressed by all somatic cells at varying levels. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesised antigenic peptides to CD8+ T lymphocytes, which are usually cytotoxic T cells. Fibroblasts or neurons however only show a low level of antigen.
Type: Primary
Antigen: MHC class 1
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité