Anti-BATF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-7463R-FITC
BOSSBS-7463R-FITCEA 900 EUR InStock
BOSSBS-7463R-FITC
Anti-BATF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
B-ATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of B-ATF mediates dimerization with members of the Jun family of proteins. The B-ATF protein does not homodimerize efficiently, but rather forms a heterodimer preferentially with c-Jun. The B-ATF/c-Jun protein complex can interact with DNA containing a consensus binding site for AP-1, suggesting that B-ATF functions as a tissue-specific modulator of the AP-1 transcription complex in human cells. B-ATF also associates with IFP35, a leucine zipper protein thatMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-7463R-FITC
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité